CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
110634490
AG110634490EA 3242 USD
AG110634490
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Mark VI spray chamber for use with early model Agilent AA instruments.
  • Simple-to-use twist and lock assembly makes for easy cleaning and routine maintenance
  • Flexible operation with the capability to tune performance using the externally adjustable impact bead
  • Fit mixing paddles for better precision and reduced burner blockage with the nitrous-oxide flame
  • When fully optimized, it provides long operating times even when analyzing solutions with high levels of dissolved solids in a fuel-rich nitrous oxide-acetylene flame
  • Can be adjusted for maximum
More Product Information