Automatic Liquid Sampler Kit
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
07673-60840
AG07673-60840KT 1576 USD
AG07673-60840
Automatic Liquid Sampler Kit
Samplers Fluid Samplers
Basic supplies kit (7673A), includes six 10 μl 23/42/cone syringes, 4 ml vial with diffusion cap (144/pack), 2 ml screw-top vials with caps (1000/pack), inlet septa (100/pack), vial rack (5/pack).