Solvent Reservoir and Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
0100-2637
101397-778EA 95.33 USD
101397-778 101401-000 101401-520 101409-916 101471-586 101472-110 101479-478 101481-788 101481-944 101483-812 101484-062 101484-064 101484-462 102963-360 102963-504 102972-154 103577-260 103579-436 103579-438 103579-440 103579-442 103579-444 103579-446 103579-448 103580-262 103580-264 103580-266 103583-196 103584-316 AG0100-2637
Solvent Reservoir and Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of precision-engineered solvent reservoirs and supplies, designed to provide high levels of sample security and longevity in LC applications.
  • Bottles have an ergonomic design, the slim profile and ergonomically shaped gripping zones on both sides of the bottle promote easier and safer use
  • A predefined labeling area with self-adhesive labels and colorful silicone bottle tags, mean that you can clearly and unambiguously identify solvent compositions
  • An easy to read durable white ceramic print layout clearly indicates the nominal volume of the bottle and allows you to quickly determine the rest contents of the bottle
  • Bottles have a GL45 thread and are compatible with stay safe caps
1 and 2 L solvent reservoirs are available in amber and in clear, for situations where sample visibility is important. Bottle head assemblies are available for both tapered and screw-top bottles. The range includes bottle caps with 3-hole inserts, bottle head assemblies with tubing and filters for capillary and nano-systems, and bottle head assemblies with connections for prep systems. Solvent bottles have been redesigned for easier use and safer handling, lowering the risk of accidents in the lab. These redesigned solvent bottles are compatible with your InfinityLab LC series instruments, and can be incorporated seamlessly into your LC workflow.