Anti-IgG Rabbit Polyclonal Antibody (Alkaline Phosphatase)

Supplier: Abcam
AB97127-1MG
ABCA_AB97127-1MGEA 332 USD
ABCA_AB97127-1MG
Anti-IgG Rabbit Polyclonal Antibody (Alkaline Phosphatase)
Antibodies
Rabbit Anti-Sheep IgG H&L (Alkaline Phosphatase).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alkaline Phosphatase
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR