Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)

Supplier: Abcam
AB96942-500UG
ABCA_AB96942-500UGEA 332 USD
ABCA_AB96942-500UG
Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)
Antibodies
Donkey Anti-Sheep IgG H&L (DyLight® 650).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR