Anti-CD10 Mouse Monoclonal Antibody [clone: CB-CALLA] (PE (Phycoerythrin)/Cy7®)

Supplier: Abcam
AB95544-50TEST
ABCA_AB95544-50TESEA 671 USD
ABCA_AB95544-50TES
Anti-CD10 Mouse Monoclonal Antibody [clone: CB-CALLA] (PE (Phycoerythrin)/Cy7®)
Antibodies
PE/Cy7® Mouse monoclonal [CB-CALLA] to CD10.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: CB-CALLA
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR