Anti-IgY Mouse Monoclonal Antibody [clone: 7C2]

Supplier: Abcam
AB8922-250UL
ABCA_AB8922-250ULEA 276 USD
ABCA_AB8922-250UL
Anti-IgY Mouse Monoclonal Antibody [clone: 7C2]
Antibodies
Mouse monoclonal [7C2] Anti-Chicken IgY H&L.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgY
Clonality: Monoclonal
Clone: 7C2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR