Anti-EPCR/CD201 Rat Monoclonal Antibody [clone: RCR-252]

Supplier: Abcam
AB81712-50UG
ABCA_AB81712-50UGEA 658 USD
ABCA_AB81712-50UG
Anti-EPCR/CD201 Rat Monoclonal Antibody [clone: RCR-252]
Antibodies
Rat monoclonal [RCR-252] to EPCR/CD201.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EPCR/CD201
Clonality: Monoclonal
Clone: RCR-252
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR