Anti-Factor IX/PTC Rabbit Polyclonal Antibody (Biotinylated)

Supplier: Abcam
AB79931-150UG
ABCA_AB79931-150UGEA 671 USD
ABCA_AB79931-150UG
Anti-Factor IX/PTC Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Biotin Rabbit polyclonal to Factor IX/PTC.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Factor IX/PTC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR