Anti-CD147 Mouse Monoclonal Antibody [clone: MEM-M6/1] (PE (Phycpoerythrin))

Supplier: Abcam
AB77133-50TEST
ABCA_AB77133-50TESEA 326 USD
ABCA_AB77133-50TES
Anti-CD147 Mouse Monoclonal Antibody [clone: MEM-M6/1] (PE (Phycpoerythrin))
Antibodies
PE Mouse monoclonal [MEM-M6/1] to CD147.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: MEM-M6/1
Conjugation: PE (Phycpoerythrin)
Epitope: ab77133 recognizes an epitope in the N terminal Ig domain (D1) of CD147
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR