Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Mouse Monoclonal Antibody [clone: 4H8]

Supplier: Abcam
AB5408-100UG
ABCA_AB5408-100UGEA 501 USD
ABCA_AB5408-100UG
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Mouse Monoclonal Antibody [clone: 4H8]
Antibodies
Mouse monoclonal [4H8] to RNA polymerase II CTD repeat YSPTSPS (phospho S5) - ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Monoclonal
Clone: 4H8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR