Anti-Metabotropic Glutamate Receptor 4/MGLUR4 Rabbit Polyclonal Antibody

Supplier: Abcam
AB53088-100UG
ABCA_AB53088-100UGEA 733 USD
ABCA_AB53088-100UG
Anti-Metabotropic Glutamate Receptor 4/MGLUR4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Metabotropic Glutamate Receptor 4/MGLUR4.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Metabotropic Glutamate Receptor 4/MGLUR4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Cat;Mouse;Rat;Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR