Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Polyclonal Antibody

Supplier: Abcam
AB5131-50UG
ABCA_AB5131-50UGEA 645 USD
ABCA_AB5131-50UG
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to RNA polymerase II CTD repeat YSPTSPS (phospho S5).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human;Saccharomyces cerevisiae;C. elegans;Drosophila melanogaster;S. pombe;Zebrafish;Xenopus laevis;A. thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR