Anti-IgG1, kappa Mouse Monoclonal Antibody [clone: 15H6] (APC/Cy7®)

Supplier: Abcam
AB46739-50TEST
ABCA_AB46739-50TESEA 326 USD
ABCA_AB46739-50TES
Anti-IgG1, kappa Mouse Monoclonal Antibody [clone: 15H6] (APC/Cy7®)
Antibodies
APC/Cy7® Mouse IgG1, kappa monoclonal [15H6] - Isotype control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: IgG1, kappa
Clonality: Monoclonal
Clone: 15H6
Conjugation: APC/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR