Human IL-12 p70 ELISA Kit
Supplier: Abcam
AB46143-1X96TESTS AB46143-2X96TESTS
ABCA_AB46143-1X96TEA 953 USD
ABCA_AB46143-1X96T ABCA_AB46143-2X96T
Human IL-12 p70 ELISA Kit
Assays ELISAs
Human IL-12 p70 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-12 p70 in Human in plasma, cell culture supernatant, serum samples.