Anti-CANX Rabbit Monoclonal Antibody [clone: EPR3632] (APC (Allophycocyanin))

Supplier: Abcam
AB319320-100UL
ABCA_AB319320-100UEA 790 USD
ABCA_AB319320-100U
Anti-CANX Rabbit Monoclonal Antibody [clone: EPR3632] (APC (Allophycocyanin))
Antibodies
APC Rabbit monoclonal [EPR3632] to Calnexin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CANX
Clonality: Monoclonal
Clone: EPR3632
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR