Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: Abcam
AB315191-100UL
ABCA_AB315191-100UEA 727 USD
ABCA_AB315191-100U
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]
Antibodies
Mouse monoclonal [3F9] to CRISPR-Cas9.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: 3F9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: S. pyogenes
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR