Anti-RNA Helicase A Rabbit Monoclonal Antibody [clone: EPR13521] (HRP (Horseradish Peroxidase))

Supplier: Abcam
AB306020-100UL
ABCA_AB306020-100UEA 708 USD
ABCA_AB306020-100U
Anti-RNA Helicase A Rabbit Monoclonal Antibody [clone: EPR13521] (HRP (Horseradish Peroxidase))
Antibodies
HRP Rabbit monoclonal [EPR13521] to RNA Helicase A.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RNA Helicase A
Clonality: Monoclonal
Clone: EPR13521
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR