Anti-SARS spike glycoprotein+COVID-19 Spike Protein Human Monoclonal Antibody [clone: CR3022]

Supplier: Abcam
AB286179-200UG
ABCA_AB286179-200UEA 852 USD
ABCA_AB286179-200U
Anti-SARS spike glycoprotein+COVID-19 Spike Protein Human Monoclonal Antibody [clone: CR3022]
Antibodies
Human monoclonal [CR3022] to SARS spike glycoprotein+COVID-19 Spike Protein.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS spike glycoprotein+COVID-19 Spike Protein
Clonality: Monoclonal
Clone: CR3022
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR