Anti-Anti-CRISPR protein AcrIIA4 Mouse Monoclonal Antibody [clone: 7C5-3A8]

Supplier: Abcam
AB273439-1MG AB273439-20UG AB273439-100UG
ABCA_AB273439-1MGEA 7840 USD
ABCA_AB273439-1MG ABCA_AB273439-20UG ABCA_AB273439-100U
Anti-Anti-CRISPR protein AcrIIA4 Mouse Monoclonal Antibody [clone: 7C5-3A8]
Antibodies
Mouse monoclonal [7C5-3A8] to Anti-CRISPR protein AcrIIA4.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Anti-CRISPR protein AcrIIA4
Clonality: Monoclonal
Clone: 7C5-3A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR