Anti-CD10 Mouse Monoclonal Antibody [clone: MME/1870]

Supplier: Abcam
AB237865-100UG
ABCA_AB237865-100UEA 1134 USD
ABCA_AB237865-100U
Anti-CD10 Mouse Monoclonal Antibody [clone: MME/1870]
Antibodies
Mouse monoclonal [MME/1870] to CD10 - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: MME/1870
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR