Human VTA1 ELISA Kit
Supplier: Abcam
AB222878-1X96TESTS
ABCA_AB222878-1X96EA 996 USD
ABCA_AB222878-1X96
Human VTA1 ELISA Kit
Assays ELISAs
Human VTA1 ELISA Kit is a sandwich (quantitative) ELISA kit for the measurement of Human VTA1 in Human in plasma, cell culture supernatant, serum samples.