Anti-IL3RA/CD123 Mouse Monoclonal Antibody [clone: 6H6] (Biotinylated)

Supplier: Abcam
AB21560-50UG
ABCA_AB21560-50UGEA 671 USD
ABCA_AB21560-50UG
Anti-IL3RA/CD123 Mouse Monoclonal Antibody [clone: 6H6] (Biotinylated)
Antibodies
Biotin Mouse monoclonal [6H6] to IL3RA/CD123.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL3RA/CD123
Clonality: Monoclonal
Clone: 6H6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR