Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody

Supplier: Abcam
AB20745-100UL
ABCA_AB20745-100ULEA 671 USD
ABCA_AB20745-100UL
Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody
Antibodies
Goat polyclonal to Respiratory Syncytial Virus.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Respiratory syncytial virus (RSV)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR