Anti-DDDDK epitope tag Rabbit Monoclonal Antibody [clone: EPR20018-251]

Supplier: Abcam
AB205606-1ML AB205606-100UL
ABCA_AB205606-1MLEA 4512 USD
ABCA_AB205606-1ML ABCA_AB205606-100U
Anti-DDDDK epitope tag Rabbit Monoclonal Antibody [clone: EPR20018-251]
Antibodies
Rabbit monoclonal [EPR20018-251] to DDDDK tag (Binds to FLAG® tag sequence).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DDDDK epitope tag
Clonality: Monoclonal
Clone: EPR20018-251
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR