Anti-Influenza A Virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
AB20388-500UL
ABCA_AB20388-500ULEA 595 USD
ABCA_AB20388-500UL
Anti-Influenza A Virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC Goat polyclonal to Influenza A Virus.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Influenza A Virus
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Influenza A
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR