Anti-Neuron navigator 1 Rat Monoclonal Antibody [clone: KT124]

Supplier: Abcam
AB201920-100UG
ABCA_AB201920-100UEA 639 USD
ABCA_AB201920-100U
Anti-Neuron navigator 1 Rat Monoclonal Antibody [clone: KT124]
Antibodies
Rat monoclonal [KT124] to Neuron navigator 1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Neuron navigator 1
Clonality: Monoclonal
Clone: KT124
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR