Anti-Milk Fat Globule 1 Rabbit Polyclonal Antibody

Supplier: Abcam
AB200649-100UL
ABCA_AB200649-100UEA 671 USD
ABCA_AB200649-100U
Anti-Milk Fat Globule 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Milk Fat Globule 1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Milk Fat Globule 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR