Anti-IgG2b Mouse Antibody [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
AB18419-50UG
ABCA_AB18419-50UGEA 182 USD
ABCA_AB18419-50UG
Anti-IgG2b Mouse Antibody [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC Mouse IgG2b, kappa [MPC-11] - isotype control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Control
Antigen: IgG2b
Clonality:
Clone: MPC-11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR