Immunoassay Blocking Buffer
Supplier: Abcam
AB171534-1000ML
AB171534-50ML
ABCA_AB171534-1000EA
495
USD
ABCA_AB171534-1000
ABCA_AB171534-50ML
Immunoassay Blocking Buffer
Buffers
Biological Buffers
Immunoassay Blocking Buffer effectively preserves the conformation and activity of dried proteins in immunoassays.
Caution:
RUO.