Anti-RFX3 Mouse Polyclonal Antibody

Supplier: Abcam
AB168475-50UG
ABCA_AB168475-50UGEA 671 USD
ABCA_AB168475-50UG
Anti-RFX3 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal to RFX3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RFX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR