Anti-Proteasome 26S S3/PSMD3 Rabbit Polyclonal Antibody

Supplier: Abcam
AB140440-100UG
ABCA_AB140440-100UEA 683 USD
ABCA_AB140440-100U
Anti-Proteasome 26S S3/PSMD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Proteasome 26S S3/PSMD3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Proteasome 26S S3/PSMD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR