Anti-beta Amyloid Chicken Polyclonal Antibody

Supplier: Abcam
AB134022-50UG
ABCA_AB134022-50UGEA 671 USD
ABCA_AB134022-50UG
Anti-beta Amyloid Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal to beta Amyloid.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: beta Amyloid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR