Anti-Spectrin beta 5 Rabbit Polyclonal Antibody

Supplier: Abcam
AB111223-200UL
ABCA_AB111223-200UEA 658 USD
ABCA_AB111223-200U
Anti-Spectrin beta 5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Spectrin beta 5.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Spectrin beta 5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR