Lactic acid 80-85% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

L14259-0B L14259-30
AAAL14259-0EEA 236.27 USD
AAAL14259-0E AAL14259-0B AAL14259-30
Lactic acid 80-85% in aqueous solution
Lactic acid

CAS No.: 50-21-5 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 110°(230°F) Refractive Index: 1.4230 Boiling Point: 122°/15mm Merck Reference: 14,5336 MDL No.: MFCD00004520

Formula: H₃CCH(OH)COOH
Boiling Pt: 122 °C/15 mm
Density: 1.209
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Merck Index: 14,5336

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR