Potassium hydride 30% w/w mineral oil

Supplier: Thermo Scientific Chemicals
Danger

L13266-ND
AAL13266-NDEA 721.46 USD
AAL13266-ND
Potassium hydride 30% w/w mineral oil
Potassium hydride

Fieser: 1,935 12,407 13,257 14,265 17,290 18,299 19,276 21,360


CAS No.: 7693-26-7 Molecular Formula: KH Formula Weight: 40.11 Flash Point: 160°(320°F) Odor: Odorless UN No.: 1409 MDL No.: MFCD00011357

Formula: HK
MW: 40.106 g/mol
Density: 1.45
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011357
CAS Number: 7693-26-7
EINECS: 231-704-8
UN: 1409
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR