(S)-(+)-α-Methoxyphenylacetic acid 99%
Supplier: Thermo Scientific Chemicals
L08748-03
L08748-MD
AAL08748-03EA
87
USD
AAL08748-03
AAL08748-MD
(S)-(+)-α-Methoxyphenylacetic acid 99%
(S)-(+)-α-Methoxyphenylacetic acid
Purity: 99%
CAS No.: 26164-26-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 63-68°
Boiling Point: 164-166°/18mm
MDL No.: MFCD00064216
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...