Sodium dodecyl sulfate (SDS) 99% for molecular biology
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade. Solid.
Purity: 99%
CAS No.: 151-21-3
Molecular Formula: C12H25NaO4S
Formula Weight: 288.38
Physical Form: Solid
Appearance: White
Melting Point: ca 206°
Solubility: Soluble in water
Beilstein Reference: 3599286
UN No.: 2926
MDL No.: MFCD00036175
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: ∼ 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,8636 |
|
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...