Anisomycin 10 mg/ml in DMSO, sterile filtered

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

J67498-XF
AAJ67498-XFEA 244.01 USD
AAJ67498-XF
Anisomycin 10 mg/ml in DMSO, sterile filtered
Anisomycin

Anisomycin is applied as an antiparasitic agent and antineoplastic agent.


Liquid


It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD). Activates mitogen-activated protein (MAP) kinases (JNK/SAPK and p38/RK).


Fully miscible in water, soluble in ethanol (50 mM), DMSO (100 mM), methanol (20 mg/mL clear, faint yellow solution), slightly soluble in alcohols, esters, ketones, chloroform, benzene, toluene, hexane, and ethyl acetate (10 mg/mL clear, colorless solution), DMF (20 mg/mL), and PBS pH 7.2 (0.5 mg/mL)


Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077650
CAS Number: 22862-76-6
EINECS: 245-269-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR