Gentamicin sulfate 20 mg/mL in water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

J67488-AD
AAJ67488-ADEA 359.84 USD
AAJ67488-AD
Gentamicin sulfate 20 mg/mL in water, sterile filtered
Gentamicin sulphate

Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered


CAS No.: 1405-41-0 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD00270181

Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR