QNZ ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline, N4-(4-Phenoxyphenethyl)quinazoline-4,6-diamine
J67487-MCR
AAJ67487-EXDEA
40.34
USD
AAJ67487-EXD
AAJ67487-LB0
AAJ67487-MCR
QNZ ≥98%
QNZ
Solid
Purity: 98%
CAS No.: 545380-34-5
Molecular Formula: C22H20N4O
Formula Weight: 356.43
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD06411436
Formula:
C₂₂H₂₀N₄O MW: 356.429 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06411436 CAS Number: 545380-34-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...