Bafilomycin B1 0.1 mg/ml in methanol, liquid, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

J67463-XF
AAJ67463-XFEA 314 USD
AAJ67463-XF
Bafilomycin B1 0.1 mg/ml in methanol, liquid, sterile filtered
Bafilomycin B1

Bafilomycin B1, 0.1 mg/ml in methanol, sterile-filtered


Liquid


Caution: Reproductive Harm

Formula: C₄₄H₆₅NO₁₃
Storage Temperature: Refrigerator
MDL Number: MFCD00242671
CAS Number: 88899-56-3
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR