Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 90 nm OD50 (Abs. 564 nm)
Supplier: Thermo Scientific Chemicals
J67449-AMI
AAJ67449-AMIEA
267
USD
AAJ67449-AMI
AAJ67449-XF
Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 90 nm OD50 (Abs. 564 nm)
Gold
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Red to pink
MDL No.: MFCD00003436
Formula:
Au Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...