Gold 99% supplied in 0.1 mM PBS, nanoparticles 5 nm reactant-free OD1 (Abs. 515-520 nm)

Supplier: Thermo Scientific Chemicals

J67448-AC J67448-AE
AAJ67448-ACEA 115 USD
AAJ67448-AC AAJ67448-AE
Gold 99% supplied in 0.1 mM PBS, nanoparticles 5 nm reactant-free OD1 (Abs. 515-520 nm)
Gold

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications


Liquid

Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR