Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 50 nm OD1 (Abs. 535 nm)

Supplier: Thermo Scientific Chemicals

J67433-AC J67433-AE
AAJ67433-ACEA 95.3 USD
AAJ67433-AC AAJ67433-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 50 nm OD1 (Abs. 535 nm)
Gold

For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.


Liquid


Applications: For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR