Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 5 nm OD50 (Abs. 515-520 nm)

Supplier: Thermo Scientific Chemicals

J67413-AMI J67413-XF
AAJ67413-AMIEA 348 USD
AAJ67413-AMI AAJ67413-XF
Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 5 nm OD50 (Abs. 515-520 nm)
Gold

Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Dark red to brown MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR