Cephalexin 50 mg/mL in ammonium hydroxide, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
J67377-AC
AAJ67377-ACEA
93.2
USD
AAJ67377-AC
AAJ67377-AD
Cephalexin 50 mg/mL in ammonium hydroxide, sterile filtered
Cephalexin
Concentration: 50 mg/ml
CAS No.: 15686-71-2
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00167148
Formula:
C₁₆H₁₉N₃O₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00167148 CAS Number: 15686-71-2 EINECS: 239-773-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...