Hygromycin B 50 mg/ml in acetic acid, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

J67371-XF
AAJ67371-XFEA 199.17 USD
AAJ67371-XF
Hygromycin B 50 mg/ml in acetic acid, sterile filtered
Hygromycin B

Hygromycin B, 50 mg/mL in distilled water, sterile-filtered.


  • Liquid


Hygromycin B was used for hygro resistance effect in the following cells transfected 293T cells with mixture of plasmids at 160 g/mL, MCF-7ALOX12 cells at 150 g/mL, MSCV Luciferase PGK-hygro plasmid and STAT3 Reporter HeLa Stable Cell Line (stably transfected with Luciferase gene under the control of STAT3 promoter) maintained in DMEM supplemented with 10% FBS.


Hygromycin B is suitable for the selection of cells transfected with Escherichia coli hygromycin resistance gene (hyg or hph), as well as for the maintenance of the Hygromycin phenotype of resistant cells. Consistent with its more specific effect on translocation, hygromycin B exhibits monophasic inhibition of polypeptide synthesis, suggesting that it binds to a single site on the ribosome.


Fully miscible in water and soluble in methanol or ethanol and buffer solution


Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR