Ethidium bromide 98% for molecular biology
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
J67270-14
AAJ67270-14EA
613
USD
AAJ67270-14
Ethidium bromide 98% for molecular biology
Ethidium bromide
Powder
Purity: 98%
CAS No.: 1239-45-8
Molecular Formula: C21H20BrN3
Formula Weight: 394.32
Physical Form: Powder
Appearance: Red to purple
Odor: Weak
Melting Point: ca 261° dec.
Beilstein Reference: 3642536
UN No.: 2811
MDL No.: MFCD00011724
Formula:
C₂₁H₂₀BrN₃ MW: 394.31 g/mol Melting Pt: ∼ 261 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011724 CAS Number: 1239-45-8 EINECS: 214-984-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...