Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 10 nm OD1 (Abs. 520 nm)

Supplier: Thermo Scientific Chemicals

J67188-AC J67188-AE
AAJ67188-ACEA 95.5 USD
AAJ67188-AC AAJ67188-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 10 nm OD1 (Abs. 520 nm)
Gold

For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.


Liquid


Applications: For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR